Methylthymol blue sodium salt, water soluble, pure

Synonyms: Sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetateMethylthymol Blue tetrasodium salt

AC10637-0050
200013-378EA 85.5 USD
200013-378
Methylthymol blue sodium salt, water soluble, pure
Methylthymol Blue sodium salt
Formula: C₃₇H₄₀N₂Na₄O₁₃S
MW: 844.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00148905
CAS Number: 1945-77-3
EINECS: 217-743-3

Order Now

Specification Test Results

Appearance Dark green to red-brown to black Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (110 °C, 1 h)
UV Lambda max 435 to 443 nm (in water)
Specific absorbance A (1%/1cm) ≥120 (at lambda max, in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR