4-Phenylbenzoic acid 99%

Warning

Synonyms: [1,1'-Biphenyl]-4-carboxylic acidBiphenyl-4-carboxylic acid4-Biphenylcarboxylic acid

AC10628-5000 AC10628-0250 AC10628-1000
103508-462EA 529 USD
103508-462 200010-222 200035-718
4-Phenylbenzoic acid 99%
4-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Melting Pt: 223…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00002553
CAS Number: 92-92-2
EINECS: 202-203-1

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 223 to 228 °C
HPLC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR