4-Phenylbenzaldehyde 99%
|
Warning
|
Synonyms:
4-Biphenylcarboxaldehyde
, p-Biphenylylaldehyde
, [1,1'-Biphenyl]-4-carbaldehyde
, 4-Biphenylcarbaldehyde
, 4-Biphenylylaldehyde
, Biphenyl-4-carboxaldehyde
, Biphenyl-4-carbaldehyde
AC10626-0050
AC10626-0250
AC10626-1000
103508-460EA
296
USD
103508-460
200023-616
200023-614
4-Phenylbenzaldehyde 99%
4-Phenylbenzaldehyde
Formula:
C₁₃H₁₀O MW: 182.22 g/mol Boiling Pt: 184 °C (11 mmHg) Melting Pt: 57…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006947 CAS Number: 3218-36-8 EINECS: 221-742-3 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 57 to 60 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...