Benzoyl chloride 99%, pure

Danger

AC10575-0025 AC10575-0010
200000-082EA 62 USD
200000-082 200000-084
Benzoyl chloride 99%, pure
Benzoyl chloride
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 197.2 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.21 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000653
CAS Number: 98-88-4
EINECS: 202-710-8
UN: 1736
ADR: 8,II
Merck Index: 13,01113

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5540 (20 °C, 589 nm)
GC ≥98.5 %
Color scale ≤40 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR