p-Benzoquinone 99%

Danger

Synonyms: Quinone1,4-Benzoquinone

AC10563-0051 AC10563-0010 AC10563-1000 AC10563-0250 AC10563-5000
103524-668EA 603 USD
103524-668 103524-672 200026-346 103524-670 200013-900
p-Benzoquinone 99%
p-Benzoquinone
Formula: C₆H₄O₂
MW: 108.1 g/mol
Melting Pt: 112…116 °C
Density: 1.31 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00001591
CAS Number: 106-51-4
EINECS: 203-405-2
UN: 2587
ADR: 6.1,II
Merck Index: 12,08259
https://us.vwr.com/stibo/web/std.lang.all/69/87/2296987.jpg

Order Now

Specification Test Results

Appearance Yellow to green Powder
Infrared spectrum Conforms
Melting point 112 to 116 °C
Titration Iodometric ≥98.5 %
Water ≤0.5 %
Specific optical rotation -218° to -224° (20 °C, 589 nm) (c=2, 1M HCl) (On dried substance)
Titration 98.5 to 101.0 % (On dried substance)
Sulfates ≤300 ppm
Chlorides ≤200 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Ninhydrin-positive substances Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR