Benzo[ghi]perylene 98+%

Warning

AC10555-1000 AC10555-0250 AC10555-0050
200018-236EA 340 USD
200018-236 103508-416 103508-414
Benzo[ghi]perylene 98+%
Benzo[ghi]perylene
Formula: C₂₂H₁₂
MW: 276.34 g/mol
Boiling Pt: >500 °C (760 mmHg)
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00004135
CAS Number: 191-24-2
EINECS: 205-883-8

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 276 to 280 °C
GC-MS ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR