Benzil 99+%

Warning

Synonyms: 1,2-Diphenyl-1,2-ethanedione

AC10541-1000 AC10541-0025 AC10541-5000
200000-068EA 26.8 USD
200000-068 103508-408 200000-070
Benzil 99+%
Benzil
Formula: (C₆H₅)C(O)C(O)(C₆H₅)
MW: 210.23 g/mol
Boiling Pt: 346…348 °C (760 mmHg)
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00003080
CAS Number: 134-81-6
EINECS: 205-157-0
Merck Index: 13,01078

Order Now

Specification Test Results

Appearance Yellow Powder or crystals
Infrared spectrum Conforms
Melting point 93 to 97 °C
GC ≥99.0 %
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤500 ppm
Potassium (K) ≤200 ppm
Sodium (Na) ≤500 ppm
Carbon compounds (as C) ≤0.01 %
Insoluble matter ≤0.02 % (in dilute HCl)
Nitrogen compounds (as N) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR