4-(Hydroxymethyl)benzyl alcohol 99%

Synonyms: 1,4-Bis(hydroxymethyl)-benzenep-Xylene-α,α'-diolp-Phenylenedimethanol1,4-Benzenedimethanol

AC10528-0500 AC10528-0100
200021-670EA 264 USD
200021-670 200053-690
4-(Hydroxymethyl)benzyl alcohol 99%
4-(Hydroxymethyl)benzyl alcohol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 138…143 °C (1 mmHg)
Melting Pt: 116…119 °C
MDL Number: MFCD00004665
CAS Number: 589-29-7
EINECS: 209-641-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 116 to 119 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR