Benzamidine hydrochloride hydrate 98%

Warning

Synonyms: Phenylamidine hydrochloride hydrateBenzenecarboximidamide hydrochloride hydrateBenzamidinium chloride hydrateBenzene-1-carboximidamide hydrochloride hydrate

AC10524-5000 AC10524-0050 AC10524-1000 AC10524-0250
200011-312EA 208 USD
200011-312 103508-892 200011-314 103508-894
Benzamidine hydrochloride hydrate 98%
Benzamidine hydrochloride hydrate
Formula: C₆H₅C(=NH)NH₂·HCl·H₂O
MW: 174.63 g/mol
Melting Pt: 80…86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066285
CAS Number: 206752-36-5
EINECS: 216-795-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 % (on dry substance)
Water ≤19 % (K.F.)
Molar absorption coefficient ≥45000 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR