Azulene 99%

Synonyms: Bicyclo[5.3.0]decapentaene

AC10518-0010 AC10518-1000
200011-278EA 197 USD
200011-278 200011-276
Azulene 99%
Azulene
Formula: C₁₀H₈
MW: 128.17 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 99…101 °C
MDL Number: MFCD00003810
CAS Number: 275-51-4
EINECS: 205-993-6
ADR: 9,III
Merck Index: 13,00928

Order Now

Specification Test Results

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 101 °C
GC ≥98.5 %
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient ≥20000 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR