Phenazone 99%

Warning

Synonyms: Antipyrine2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one

AC10497-5000 AC10497-1000 AC10497-0050
200036-580EA 93.7 USD
200036-580 103508-862 200018-184
Phenazone 99%
Phenazone
Formula: C₁₁H₁₂N₂O
MW: 188.23 g/mol
Boiling Pt: 319 °C (760 mmHg)
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003146
CAS Number: 60-80-0
EINECS: 200-486-6
Merck Index: 12,00757

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 109 to 113 °C
Titration Iodimetric ≥98.5 %
Specific optical rotation +33° to +37° (20 °C, 589 nm) (c=0.6, ethanol)
Titration with HClO4 98.0 to 102.0 % (On dried basis)
Heavy metals (as Pb) ≤50 ppm
Impurity ≤2.0 % Sodium deoxycholate

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR