Anthranilamide 99+%

Warning

Synonyms: 2-Aminobenzamide

AC10490-5000 AC10490-0050 AC10490-1000
200030-442EA 28.1 USD
200030-442 200011-252 103508-854
Anthranilamide 99+%
Anthranilamide
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: 111…114 °C (dec.)
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00007981
CAS Number: 88-68-6
EINECS: 201-851-2

Order Now

Specification Test Results

Appearance Light brown to grey-brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 111 to 114 °C
HPLC ≥99.0 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.0002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR