9-Anthracenecarboxylic acid 99%

Warning

Synonyms: Anthracene-9-carboxylic acid

AC10488-0100
200018-170EA 86.5 USD
200018-170
9-Anthracenecarboxylic acid 99%
9-Anthracenecarboxylic acid
Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Boiling Pt: 411 °C (1013 hPa)
Melting Pt: 213…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00001257
CAS Number: 723-62-6
EINECS: 211-964-9

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 214 to 218 °C
Titration with TBAH ≥98.5 %
Free amine ≤0.0002 %
Acidity (CH3COOH) ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR