o-Anisidine 99+%
|
Danger
|
Synonyms:
2-Aminoanisole
, 2-Anisidine
, 2-Methoxyaniline
AC10481-0010
AC10481-2500
AC10481-0050
200000-038EA
62.2
USD
200000-038
200038-324
200020-200
o-Anisidine 99+%
o-Anisidine
Formula:
C₇H₉NO MW: 123.15 g/mol Boiling Pt: 225 °C (1013 hPa) Melting Pt: 3…6 °C Density: 1.092 g/cm³ Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007688 CAS Number: 90-04-0 EINECS: 201-963-1 UN: 2431 ADR: 6.1,III Merck Index: 13,00670 |
Specification Test Results
Appearance | Clear colorless to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5730 to 1.5755 (20 °C, 589 nm) |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...