3,4-Dimethoxyaniline 98%

Warning

AC10472-0100 AC10472-0250
200023-596EA 85.7 USD
200023-596 200010-206
3,4-Dimethoxyaniline 98%
3,4-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 174…176 °C (22 mmHg)
Melting Pt: 86…88 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008394
CAS Number: 6315-89-5
EINECS: 228-647-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 86 to 88 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR