4-Amino-4H-1,2,4-triazole 99%

Warning

Synonyms: 4-Amino-1,2,4-triazole4H-1,2,4-Triazol-4-amine1,2,4-Triazol-4-ylamine

AC10470-0250 AC10470-1000
103508-850EA 179 USD
103508-850 200010-176
4-Amino-4H-1,2,4-triazole 99%
4-Amino-4H-1,2,4-triazole
Formula: C₂H₄N₄
MW: 84.08 g/mol
Melting Pt: 84 °C
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00003099
CAS Number: 584-13-4
EINECS: 209-533-5

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥98.5 %
UV at 250 nm A: ≤0.01
UV at 300 nm A: ≤0.005
Residue after evaporation ≤2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR