3-Aminopyridine 99%

Danger

Synonyms: 3-PyridylaminePyridin-3-amine

AC10456-1000 AC10456-5000 AC10456-0250
200005-214EA 31.1 USD
200005-214 200016-946 200009-796
3-Aminopyridine 99%
3-Aminopyridine
Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: ∼248 °C (1013 hPa)
Melting Pt: 60…63 °C
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00006400
CAS Number: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: 6.1,II
Merck Index: 13,00472

Order Now

Specification Test Results

Appearance Off-white to brown Crystals or flakes or crystalline mass
Infrared spectrum Conforms
Melting point 60 to 63 °C
GC ≥98.5 %
Moisture ≤3.0 % (as packaged)
Viscosity 0.2 to 0.6 s
Free acid (CH3COOH) ≤0.06 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR