(S)-(+)-2-Amino-1-propanol 98%

Danger

Synonyms: L(+)-AlaninolL-AlaninolL(+)-2-Amino-1-propanol(S)-2-Aminopropan-1-ol

AC10445-0100 AC10445-0010 AC10445-0500
103530-178EA 972 USD
103530-178 200008-248 200015-350
(S)-(+)-2-Amino-1-propanol 98%
(S)-(+)-2-Amino-1-propanol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 173…176 °C (760 mmHg)
Density: 0.965 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS Number: 2749-11-3
EINECS: 220-388-7
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4510 (20 °C, 589 nm)
TLC ≥97.5 %
Specific optical rotation +20° to +24° (20 °C, 589 nm) (c=2, EtOH)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR