(4-Aminophenyl)acetic acid 98%

Warning

Synonyms: 4-Aminophenylacetic acid

AC10428-1000 AC10428-0250 AC10428-0050
103508-826EA 182 USD
103508-826 200032-436 200013-876
(4-Aminophenyl)acetic acid 98%
(4-Aminophenyl)acetic acid
Formula: (C₆H₄(NH₂))CH₂COOH
MW: 151.16 g/mol
Melting Pt: 196…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00007916
CAS Number: 1197-55-3
EINECS: 214-828-7
Merck Index: 13,00462

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 195 to 201 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.01 %
Iron (Fe) ≤0.002 %
Magnesium (Mg) ≤0.01 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.01 %
Strontium (Sr) ≤0.1 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 % (in dilute HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR