(±)-2-Aminoheptane 98+%

Danger

Synonyms: (±)-Tuaminoheptane2-Heptylamine(±)-1-Methylhexylamine

AC10377-1000 AC10377-5000 AC10377-0250
103524-636EA 457 USD
103524-636 200034-822 200005-066
(±)-2-Aminoheptane 98+%
(±)-2-Aminoheptane
Formula: C₇H₁₇N
MW: 115.22 g/mol
Boiling Pt: 142…144 °C (760 mmHg)
Density: 0.768 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008101
CAS Number: 123-82-0
EINECS: 204-655-5
UN: 2734
ADR: 8,II
Merck Index: 12,09934

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4175 to 1.4195 (20 °C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR