2-Amino-5-chloropyridine 98%

Warning

Synonyms: 5-Chloropyridin-2-amine5-Chloro-2-pyridylamine

AC10341-5000 AC10341-1000 AC10341-0250
200016-310EA 128 USD
200016-310 200040-816 200016-308
2-Amino-5-chloropyridine 98%
2-Amino-5-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 127…128 °C (11 mmHg)
Melting Pt: 134…138 °C
Flash Pt: 160 °C
MDL Number: MFCD00006324
CAS Number: 1072-98-6
EINECS: 214-020-4

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 134 to 138 °C
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %
Copper (Cu) ≤20 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤20 ppm
Silver (Ag) ≤10 ppm
Insoluble matter ≤0.005 % (in HNO3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR