(±)-2-Aminobutan-1-ol 97%

Danger

Synonyms: 2-Aminobutan-1-ol

AC10325-5000 AC10325-0050 AC10325-1000
200039-240EA 21.1 USD
200039-240 200015-222 200033-226
(±)-2-Aminobutan-1-ol 97%
(±)-2-Aminobutan-1-ol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.944 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008095
CAS Number: 96-20-8
EINECS: 202-488-2
UN: 2735
ADR: 8,III
Merck Index: 13,00426

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4550 (20 °C, 589 nm)
Water ≤1.5 %
GC ≥96.0 %
Specific optical rotation -60° to -64° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR