2-Aminobenzophenone 98%

Warning

Synonyms: o-Aminobenzophenone

AC10316-1000 AC10316-0250
200034-616EA 225 USD
200034-616 200009-166
2-Aminobenzophenone 98%
2-Aminobenzophenone
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 105…108 °C
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00007713
CAS Number: 2835-77-0
EINECS: 220-613-9

Order Now

Specification Test Results

Appearance Yellow Powder and/or chunks
Infrared spectrum Conforms
Melting point 105 to 108 °C
Titration with HClO4 ≥97.5 %
Titration by coupling ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR