4-Aminoantipyrine 98%

Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one4-Aminophenazone

AC10315-5000 AC10315-1000 AC10315-0250
200005-358EA 72.9 USD
200005-358 200010-184 200005-360
4-Aminoantipyrine 98%
4-Aminoantipyrine
Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 105.5…110 °C
Density: 0.8 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Order Now

Specification Test Results

Appearance Yellow to yellow-brown Powder or crystals
Infrared spectrum Conforms
Melting point 105 to 110 °C
Loss on drying ≤1.5 %
Sulfated ash ≤0.1 %
Solubility (5 % in water) Clear
Titration with HClO4 ≥97.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR