Allylbenzene 98%

Danger

Synonyms: 3-Phenylpropene1-Phenyl-2-propene

AC10289-1000 AC10289-5000 AC10289-0250
200011-026EA 61.4 USD
200011-026 103524-016 200024-326
Allylbenzene 98%
Allylbenzene
Formula: (C₆H₅)CH₂CHCH₂
MW: 118.18 g/mol
Boiling Pt: 156…157 °C (760 mmHg)
Density: 0.89 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00008651
CAS Number: 300-57-2
EINECS: 206-095-7
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5095 to 1.5135 (20 °C, 589 nm)
GC ≥97.5 %
Appearance of solution Clear colorless
Related substances ≤0.5 % (impurity A)
Related substances ≤0.1 % (impurity C)
Related substances ≤0.05 % (unspecified impurities)
Related substances ≤0.8 % (total impurities)
pH 4.0 to 6.0
Specific optical rotation +33.7° to +36.0° (20 °C, 589 nm) (on dried substance)
Titration with HClO4 99.0 to 101.0 % (on dried substance)
Sulfates ≤200 ppm
Chlorides ≤200 ppm
Ammonium (NH4) ≤0.1 %
Iron (Fe) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR