Adipic acid 99%

Danger

Synonyms: 1,6-Hexanedioic acidHexanedioc acid

AC10281-0010 AC10281-0025 AC10281-0250 AC10281-5000
200010-986EA 54.5 USD
200010-986 200000-010 103512-376 103508-478
Adipic acid 99%
Adipic acid
Formula: HOOC(CH₂)₄COOH
MW: 146.14 g/mol
Boiling Pt: 337 °C (760 mmHg)
Melting Pt: 151…153 °C
Flash Pt: 196 °C
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 13,00163
https://us.vwr.com/stibo/web/std.lang.all/02/30/2300230.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 153 °C
Water ≤0.2 %
Ash ≤0.0002 %
Titration with NaOH ≥99 %
Color of solution ≤5 APHA (20g adipic acid + 20 g dist. H2O + 60 g 10% NH3 soln.)
Nitrate (NO3) ≤4 ppm
Iron (Fe) ≤0.4 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR