Acetylthiocholine iodide 98%

Danger

Synonyms: S-Acetylthiocholine iodide(2-Mercaptoethyl)trimethylammonium iodide acetate2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide

AC10271-0050 AC10271-0250
103524-008EA 299 USD
103524-008 200007-586
Acetylthiocholine iodide 98%
Acetylthiocholine iodide
Formula: H₃CC(S)O(CH₂)₂N(CH₃)₃I
MW: 289.18 g/mol
Melting Pt: 205…210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011819
CAS Number: 1866-15-5
EINECS: 217-474-1
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/05/63/2380563.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR