Acetophenone 98%, pure

Warning

Synonyms: Phenyl methyl ketoneMethyl phenyl ketone

AC10241-0100 AC10241-2500
200000-006EA 23.3 USD
200000-006 103508-510
Acetophenone 98%, pure
Acetophenone
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 202 °C (760 mmHg)
Melting Pt: 19.6 °C
Density: 1.028 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
ADR: 9,
Merck Index: 13,00074

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5350 (20 °C, 589 nm)
GC ≥97.5 %
Visual transition interval from pH 3.5 (yellow) to pH 6.5 (pink)
Visual transition interval from pH 9.4 (orange-brown) to pH 12.0 (violet)
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR