Acetyl chloride for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.22252.0100
CA8.22252.0100
Acetyl chloride for synthesis, Sigma-Aldrich®
Acetyl chloride

8.22252.0100 replaces and is the same as AX0185-1. Part # Change only, Same Pack Size, Same Specifications.

Formula: C₂H₃ClO
MW: 78.5 g/mol
Boiling Pt: 50.9 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3 (8),II
Merck Index: 13,00087

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR