Sodium cyanotrihydroborate ≥95% for synthesis, Sigma-Aldrich®

Danger

Synonyms: Sodium cyanoborohydride

M-Clarity™ quality level = MQ200

8.18053.0500 8.18053.0025
EM8.18053.0025EA 449 USD
EM8.18053.0025 EM8.18053.0500
Sodium cyanotrihydroborate ≥95% for synthesis, Sigma-Aldrich®
Sodium cyanotrihydroborate
Formula: CH₃BNNa
MW: 62.84 g/mol
Melting Pt: 241…243 °C
Density: 1.11…1.12 g/cm³ (28 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003516
CAS Number: 25895-60-7
EINECS: 247-317-2
UN: 3134
ADR: 4.3 (6.1),I
Merck Index: 12,08751

Order Now

Specification Test Results

Assay (iodometric) ~95% min
Identity (IR-spectrum) Conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR