m-Cresol, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 3-Hydroxytoluene3-Methylphenol1-Hydroxy-3-methylbenzene

M-Clarity™ quality level = MQ200

8.09691.9030 8.09691.9030
EM8.09691.9030EA 1377 USD
EM8.09691.9030 CA8.09691.9030
m-Cresol, Sigma-Aldrich®
m-Cresol
Formula: CH₃C₆H₄OH
MW: 108.14 g/mol
Boiling Pt: 202.2 °C (1013 hPa)
Melting Pt: 11.5 °C
Density: 1.0336 g/cm³ (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002302
CAS Number: 108-39-4
EINECS: 203-577-9
UN: 2076
ADR: 6.1,II
Merck Index: 13,02604
https://us.vwr.com/stibo/web/std.lang.all/08/21/2300821.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR