Cumene, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-PhenylpropaneIsopropylbenzene

M-Clarity™ quality level = MQ200

8.02681.1000 8.02681.1000
EM8.02681.1000EA 64 USD
CA8.02681.1000 EM8.02681.1000
Cumene, Sigma-Aldrich®
Cumene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –96 °C
Density: 0.8624 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00008881
CAS Number: 98-82-8
EINECS: 202-704-5
UN: 1918
ADR: 3,III
Merck Index: 13,02644

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR