Quinoline, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.02407.0250
8.02407.0250
EM8.02407.0250EA
90
USD
EM8.02407.0250
CA8.02407.0250
Quinoline, Sigma-Aldrich®
Quinoline
Formula:
C₉H₇N MW: 129.16 g/mol Boiling Pt: 238 °C (1013 hPa) Melting Pt: –15 °C Density: 1.095 g/cm³ (20 °C) Flash Pt: 107 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00006736 CAS Number: 91-22-5 EINECS: 202-051-6 UN: 2656 ADR: 6.1,III Merck Index: 12,08253 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...