KIMBERLY-CLARK® Scrub Pant, KIMBERLY-CLARK PROFESSIONAL®
69714 69712 69713 69711 69711 69714 69712 69713
94001-638CS 133.92 USD
94001-638 89135-418 89135-420 94001-636 94001-642 89135-424 89135-422 94001-640
KIMBERLY-CLARK® Scrub Pant, KIMBERLY-CLARK PROFESSIONAL®
Single-use scrub pants provide a comfortable and effective alternative to the losses, laundry expenses, and replacement costs associated with reusable garments.
  • Protective Fabric
  • Comfortable Use for Extended Wear
  • Single Use
Scrub pants are made of a soft, protective fabric designed to shield the wearer from light fluid contact
These generously sized items allow for comfortable use during extended wear. All pants feature waist ties, reinforced side and rear pockets, and reinforced in-seams.
This fabric is manufactured from a protective, three-layer spunbond-meltblown-spunbond (SMS) fabric.