3,4,5-Trifluorocinnamic acid 97+%

Warning

B23894-06 B23894-03
AAB23894-06EA 257.32 USD
AAB23894-06 AAB23894-03
3,4,5-Trifluorocinnamic acid 97+%
3,4,5-Trifluorocinnamic acid
Formula: C₉H₅F₃O₂
MW: 202.13 g/mol
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00083529
CAS Number: 152152-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR