4-Benzylpiperidine 98%

B23883-14 B23883-22 B23883-36 B23883-36 B23883-22 B23883-14
AAB23883-36EA 445.99 USD
AAB23883-36 AAB23883-14 CAAAB23883-14 CAAAB23883-36 CAAAB23883-22 AAB23883-22
4-Benzylpiperidine 98%
4-Benzylpiperidine
Formula: C₁₂H₁₇N
MW: 175.27 g/mol
Boiling Pt: 278…279 °C
Melting Pt: 6…7 °C
Density: 0.997
Flash Pt: 147 °C(296 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006006
CAS Number: 31252-42-3
EINECS: 250-535-0
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR