2-Chloro-6-nitrotoluene 98+%

Warning

B23675-0B B23675-30 B23675-0B
AAB23675-0BEA 183.74 USD
CAAAB23675-30 AAB23675-0B CAAAB23675-0B
2-Chloro-6-nitrotoluene 98+%
2-Chloro-6-nitrotoluene
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 238 °C
Melting Pt: 34…38 °C
Flash Pt: 125 °C(257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007205
CAS Number: 83-42-1
EINECS: 201-475-9
UN: 3457
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR