3-Ethylphenol 95%

Danger

Synonyms: 1-Ethyl-3-hydroxybenzene

B23470-30 B23470-30 B23470-18 B23470-09
AAB23470-30EA 591.06 USD
CAAAB23470-30 AAB23470-30 CAAAB23470-09 CAAAB23470-18
3-Ethylphenol 95%
3-Ethylphenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 218 °C
Melting Pt: –4 °C
Density: 1.028
Flash Pt: 94 °C(202 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002311
CAS Number: 620-17-7
EINECS: 210-627-3
UN: 3145
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR