α,α,α,4-Tetrachlorotoluene 98+%

Danger

Synonyms: 4-Chlorobenzotrichloride1-Chloro-4-(trichloromethyl)benzene

B23446-14 B23446-22 B23446-36 B23446-14 B23446-22 B23446-36
AAB23446-14EA 29.87 USD
CAAAB23446-14 CAAAB23446-22 AAB23446-14 CAAAAB23446-36 AAB23446-22 AAAB23446-36
α,α,α,4-Tetrachlorotoluene 98+%
α,α,α,4-Tetrachlorotoluene
Formula: C₇H₄Cl₄
MW: 229.92 g/mol
Boiling Pt: 245 °C
Melting Pt: 5…6 °C
Density: 1.495
Flash Pt: 140 °C(284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000790
CAS Number: 5216-25-1
EINECS: 226-009-1
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR