1-Chlorohexadecane 97%

Supplier: Thermo Scientific Chemicals

B23406-AP B23406-AE B23406-AE
AAB23406-AEEA 37.89 USD
AAB23406-AE AAAB23406-AP CAAAB23406-AE
1-Chlorohexadecane 97%
1-Chlorohexadecane
Formula: C₁₆H₃₃Cl
MW: 260.89 g/mol
Boiling Pt: 322 °C
Melting Pt: 8…10 °C
Density: 0.865
Flash Pt: 136 °C(276 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000959
CAS Number: 4860-03-1
EINECS: 225-461-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR