4-(Trifluoromethoxy)phenylboronic acid 98%

Warning

Synonyms: Dihydroxy(4-(trifluoromethoxy)phenyl)borane(4-(Trifluoromethoxy)phenyl)metaboric acid4-Trifluoromethoxyphenylboronic acid4-(Trifluoromethoxy)benzeneboronic acid

B23233-03 B23233-14 B23233-03 B23233-06 B23233-06 B23233-14
AAB23233-06EA 158.34 USD
CAAAB23233-14 CAAAB23233-03 CAAAB23233-06 AAB23233-06 AAB23233-03 AAB23233-14
4-(Trifluoromethoxy)phenylboronic acid 98%
4-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: ca 120…125 °C
Storage Temperature: Ambient
MDL Number: MFCD01074648
CAS Number: 139301-27-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR