Bromocycloheptane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cycloheptyl bromide

B23110-06 B23110-22 B23110-14 B23110-22 B23110-06 B23110-14
AAB23110-14EA 88.35 USD
CAAAB23110-22 CAAAB23110-14 CAAAB23110-06 AAB23110-14 AAB23110-22 AAB23110-06
Bromocycloheptane 97%
Bromocycloheptane
Formula: C₇H₁₃Br
MW: 177.08 g/mol
Boiling Pt: 71…72 °C/10 mm
Density: 1.300
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004149
CAS Number: 2404-35-5
EINECS: 219-294-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR