3-Chloro-4-fluorobenzyl alcohol 97%

Warning

Synonyms: (3-Chloro-4-fluorophenyl)methanol3-Chloro-4-fluorobenzyl Alcohol

B23080-06 B23080-03 B23080-03 B23080-06
AAB23080-03EA 57.56 USD
CAAAB23080-03 AAB23080-03 AAB23080-06 CAAAB23080-06
3-Chloro-4-fluorobenzyl alcohol 97%
3-Chloro-4-fluorobenzyl alcohol
Formula: C₇H₆ClFO
MW: 160.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631550
CAS Number: 161446-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR