3-Chloro-2-fluorobenzaldehyde 97%

Warning

B22975-03 B22975-14 B22975-14 B22975-06 B22975-06 B22975-03
AAAB22975-14EA 490.12 USD
CAAAAB22975-14 AAAB22975-14 CAAAB22975-06 AAB22975-03 CAAAB22975-03 AAB22975-06
3-Chloro-2-fluorobenzaldehyde 97%
3-Chloro-2-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Density: 1,350
Storage Temperature: Ambient
MDL Number: MFCD01631571
CAS Number: 85070-48-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR