3'-Fluoroacetanilide 98%

Warning

Synonyms: N-(3-Fluorophenyl)acetamide

B22889-22 B22889-22 B22889-14 B22889-06 B22889-06 B22889-14
AAB22889-06EA 23.21 USD
AAB22889-06 AAB22889-14 CAAAB22889-14 CAAAB22889-22 AAB22889-22 CAAAB22889-06
3'-Fluoroacetanilide 98%
3'-Fluoroacetanilide
Formula: C₈H₈FNO
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00017917
CAS Number: 351-28-0
EINECS: 206-509-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR