2,3,5-Trimethylhydroquinone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3,5-Trimethylbenzene-1,4-diolTrimethylhydroquinone

B22819-36 B22819-36
AAB22819-36EA 167.99 USD
CAAAB22819-36 AAB22819-36
2,3,5-Trimethylhydroquinone 98%
2,3,5-Trimethylhydroquinone
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Melting Pt: 169…174 °C
Flash Pt: 140 °C(284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002346
CAS Number: 700-13-0
EINECS: 211-838-3
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR