3'-Fluoro-4'-(trifluoromethyl)propiophenone 97+%

Warning

B22814-03 B22814-03 B22814-06 B22814-06
AAB22814-06EA 220.89 USD
CAAAB22814-06 AAB22814-06 CAAAB22814-03 AAB22814-03
3'-Fluoro-4'-(trifluoromethyl)propiophenone 97+%
3'-Fluoro-4'-(trifluoromethyl)propiophenone
Formula: C₁₀H₈F₄O
MW: 220.17 g/mol
Melting Pt: 35…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00236306
CAS Number: 237761-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR