2-(3-Methoxyphenyl)cyclohexanone tech. 90%, Technical Grade

B22619-03 B22619-06 B22619-03 B22619-14 B22619-06 B22619-14
AAB22619-03EA 73.08 USD
CAAAB22619-03 CAAAB22619-14 AAB22619-03 AAB22619-14 AAB22619-06 CAAAB22619-06
2-(3-Methoxyphenyl)cyclohexanone tech. 90%, Technical Grade
2-(3-Methoxyphenyl)cyclohexanone
Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Boiling Pt: 140…142 °C/0.2 mm
Density: 1.100
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001630
CAS Number: 15547-89-4
EINECS: 239-602-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR