Benzoylcholine iodide 98+%

Synonyms: (2-Benzoyloxyethyl)trimethylammonium iodide

B22387-03 B22387-14 B22387-14 B22387-06 B22387-06 B22387-03
AAB22387-03EA 32.45 USD
AAB22387-03 AAB22387-14 AAB22387-06 CAAAB22387-14 CAAAB22387-03 CAAAB22387-06
Benzoylcholine iodide 98+%
Benzoylcholine iodide
Formula: C₆H₅C(O)O(CH₂)₂N(CH₃)₃I
MW: 335.18 g/mol
Melting Pt: ca 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038726
CAS Number: 17518-43-3
EINECS: 241-517-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR