2-Isobutylpyridine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22349-06
B22349-14
B22349-14
B22349-06
AAB22349-06EA
79.89
USD
AAB22349-06
AAB22349-14
CAAAB22349-06
CAAAB22349-14
2-Isobutylpyridine 99%
2-Isobutylpyridine
Formula:
C₉H₁₃N MW: 135.21 g/mol Density: 0.899 Storage Temperature: Ambient |
MDL Number:
MFCD02262142 CAS Number: 6304-24-1 EINECS: 228-606-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...